Microplate washers, automatic, Autura 1000
Supplier: MIKURA LTD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MIKU8.1000EA 4130 GBP
MIKU8.1000 MIKU24.1000 MIKU16.1000
Microplate washers, automatic, Autura 1000
Microplate Washers
The Autura 1000 range is designed for the fully automated washing and filling of both, 96 and 384 well microplates, of all possible formats.
  • Accurate dispensing precision and low residual volumes
  • Up to two wash reagents, each with its own fluid pathways
  • Open construction for easy robotic integration
  • Plate sweep for the lowest residual volumes
  • Extremely easy to clean and maintain
  • Can be built to accommodate any size of plate