Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dry ice maker, Economy Frigimat®

Supplier: Bel-Art
Frigimat®
In these dry ice makers, the epoxy-coated steel frame has four non-skid rubber feet to prevent sliding.

  • Produce a solid 454 g block of dry ice in one to two minutes
  • Each 22 ,5 kg cylinder of liquid CO₂ with a dip tube will make 8 or 9 blocks of dry ice measuring 92×104 mm
  • The PE ice chamber is held in place by a metal clamping mechanism, and a porous PE diffusion plate vents excess gas
  • With a built-in gas pressure gauge and relief valve

W×D×H: 370×140×150 mm

Shipping Weight: 3,9 kg

Delivery information: Include a 1,8 m nickel plated copper connecting tube with standard CGA 320 (0.830"-14) thread couplings for easy attachment to a LCO² cylinder.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Production of 8 - 9 blocks with 92×104 mm long 1 BELAH388760002
Production of 8 - 9 blocks with 92×104 mm long 1 BELAH388760003
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty