Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Analytical balances, Entris®

Supplier: Sartorius
Entris®
An entry level series of analytical balances, specially designed to achieve effective and reliable weighing results in routine daily work. The easy and clearly structured user interface, with its logical key assignment and excellent readability, are ideal features for safe and efficient operation.

  • Integrated basic applications: Animal weighing, density determination, counting, conversion or percentage
  • Monolithic weighing system ensures long term, high measurement accuracy
  • Linearity: ±0,2 mg, with stabilisation times typically 2,5 seconds
  • Excellent readability due to the backlit and high contrast display with 15 mm height digits
  • Draught shield with three sliding doors, easy to dismantle and clean
  • With manual leveling and manual draftshield



 

ORDER

Expand All / Collapse All

Balances with internal calibration
Model Weighing cap. Readability Pan/platform size Linearity Pk VWR Catalog Number Quantity
ENTRIS64i-1S 60 g 0,1 mg Ø 90 mm 0,2 mg 1 611-2985H
ENTRIS124i-1S 120 g 0,1 mg Ø 90 mm 0,2 mg 1 611-2981H
ENTRIS224i-1S 220 g 0,1 mg Ø 90 mm 0,2 mg 1 611-2983H

Balances with external calibration
Model Weighing cap. Readability Pan/platform size Linearity Pk VWR Catalog Number Quantity
ENTRIS64-1S 60 g 0,1 mg Ø 90 mm 0,2 mg 1 611-2984H
ENTRIS124-1S 120 g 0,1 mg Ø 90 mm 0,2 mg 1 611-2980H
ENTRIS224-1S 220 g 0,1 mg Ø 90 mm 0,2 mg 1 611-2982H
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Printer YDP20-0CE with connector cable for Entris® balances 1 611-1774
Ink ribbon cartridge for printers YDP03-0CE, YDP10-0CE and YDP20-0CE 1 611-0737
Paper rolls for printers YDP03-0CE, YDP10-0CE and YDP20-0CE, 50 metres 5 611-0738
Connection cable to a USB port 1 611-1614
Density determination kit for M-Pact and Entris® analytical balances 1 611-0909
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty