4-Chloro-6-methylquinoline ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H59890.03EA
134
GBP
H59890.03
4-Chloro-6-methylquinoline ≥97%
4-Chloro-6-methylquinoline
Formula:
C₁₀H₈ClN MW: 177.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02684204 CAS Number: 18436-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...