5-Bromo-2-isopropoxybenzonitrile ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H50627.03EA
171
GBP
H50627.03
5-Bromo-2-isopropoxybenzonitrile ≥97%
5-Bromo-2-isopropoxybenzonitrile
Formula:
C₁₀H₁₀BrNO MW: 240.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02257441 CAS Number: 515832-52-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...