Eriochrome blue black B indicator for metal titration, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
1.03168.0025EA
76.5
GBP
1.03168.0025
Eriochrome blue black B indicator for metal titration, Supelco®
Eriochrome Blue Black B
Formula:
C₂₀H₁₃N₂NaO₅S MW: 416.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003934 CAS Number: 3564-14-5 EINECS: 222-639-6 |
Specification Test Results
Solubility | 20 g/l (20 °C) |
Molar Mass | 416.39 g/mol |
Bulk Density | 540 kg/m3 |
pH | 9.5 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...