4-Chloro-3-ethoxy-2-fluoroaniline ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H33541.MDEA
156
GBP
H33541.MD
H33541.03
4-Chloro-3-ethoxy-2-fluoroaniline ≥97%
4-Chloro-3-ethoxy-2-fluoroaniline
Formula:
C₈H₉ClFNO MW: 189.61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687218 CAS Number: 1323966-31-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...