2-Chloro-6-(trifluoromethoxy)benzonitrile ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H33369.03EA
109
GBP
H33369.03
H33369.MD
2-Chloro-6-(trifluoromethoxy)benzonitrile ≥97%
2-Chloro-6-(trifluoromethoxy)benzonitrile
Formula:
C₈H₃ClF₃NO MW: 221.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18393805 CAS Number: 1261779-40-1 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...