2-Methyl-6-nitroanisole
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBAN-2890-1GEA
19.78
GBP
COBBAN-2890-1G
2-Methyl-6-nitroanisole
2-Methyl-6-nitroanisole
Formula:
C₈H₉NO₃ |
MDL Number:
MFCD00661626 CAS Number: 18102-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...