5-Hydroxy-o-toluic acid ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
5-Hydroxy-2-methylbenzoic acid
H60774.03EA
51
GBP
H60774.03
5-Hydroxy-o-toluic acid ≥97%
5-Hydroxy-o-toluic acid
Formula:
C₈H₈O₃ MW: 152.15 g/mol Melting Pt: 182…187 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09833184 CAS Number: 578-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...