4-Bromo-2-nitro-6-(trifluoromethyl)aniline ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Amino-5-bromo-3-nitrobenzotrifluoride
H61933.03EA
73.9
GBP
H61933.03
4-Bromo-2-nitro-6-(trifluoromethyl)aniline ≥98%
4-Bromo-2-nitro-6-(trifluoromethyl)aniline
Formula:
C₇H₄BrF₃N₂O₂ MW: 285.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835318 CAS Number: 157026-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...