3-Bromo-2-fluorotoluene ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Bromo-2-fluoro-3-methylbenzene
H61492.14EA
78.6
GBP
H61492.14
3-Bromo-2-fluorotoluene ≥98%
3-Bromo-2-fluorotoluene
Formula:
C₇H₆BrF MW: 189.02 g/mol Boiling Pt: 118…123 °C Melting Pt: 118…123 °C Density: 1.503 Flash Pt: 76 °C (169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04972694 CAS Number: 59907-12-9 EINECS: 261-981-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...