2-Chloro-4-fluoro-5-nitrobenzoic acid ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H61753.14EA
162
GBP
H61753.14
2-Chloro-4-fluoro-5-nitrobenzoic acid ≥97%
2-Chloro-4-fluoro-5-nitrobenzoic acid
Formula:
C₇H₃ClFNO MW: 219.55 g/mol Melting Pt: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115715 CAS Number: 114776-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...