5-Bromo-2-fluorobenzamide ≥96%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H62457.MDEA
61.5
GBP
H62457.MD
H62457.03
5-Bromo-2-fluorobenzamide ≥96%
5-Bromo-2-fluorobenzamide
Formula:
C₇H₅BrFNO MW: 218.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094430 CAS Number: 214210-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...