Rotational viscometers, VR3000
Supplier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMEDV1308EA 4300 GBP
SIMEDV1308
Rotational viscometers, VR3000
These rotational viscometers are designed for the rapid determination of viscosity.
  • (L) Low to medium viscosity
  • (R) Medium to high viscosity
  • (H) High to very high viscosity
The V1 model features a ViscosoftBasic software for data gathering and a PT100 temperature probe.
The V2 model additionnally has two more low speeds, a wider range, and a bi-directional RS232 for automated control.
Certifications: ISO 2555 − ASTM