Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.

Lateral Flow test for Bacillus cereus, Duopath® Cereus Enterotoxins

Supplier: Merck
Bacillus cereus is an environmentally widespread gram-positive, spore-forming, motile rod, which can cause gastrointestinal (e.g. diarrhoea) as well as non gastrointestinal diseases (e.g. septicemia, endocarditis, infections of the central nervous system) in humans. The illness is normally self-limiting and of short duration, although a few fatal cases have occurred. Reporting of Bacillus cereus food poisoning is not required, and as a result a low number of registered cases is regarded as representing only about 1% of actual cases. The frequency of cases also varies geographically, accounting for less than 1% of all food-related illnesses in some countries while more than 30% in others. In addition to this, some Bacillus cereus strains produce a different, heat-stable toxin known as Bacillus cereus emetic toxin. This toxin is assumed to be pre-formed in the food and is often associated with starchy products, such as rice and pasta. For these reasons, it is becoming increasingly relevant to test foods for toxin-forming Bacillus cereus using reliable and rapid testing methods. Innovative, rapid tests make screening for pathogenic organisms easier. This one-step format reduces the possibility of errors during handling.

  • Easy to use: Simply add sample and read off the result
  • Safe: Clear and distinctive positive or negative test results with built-in positive control
  • Reliable: As sensitive as the official culture media method
  • Economical: Rapid results save labour and inventory costs

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Duopath® Cereus Enterotoxins 25 Tests 1.04146.0001
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Expand All / Collapse All

Description Standard Pk Pack type VWR Catalog Number Quantity
CGYE (Caseinhydrolysate-glucose-yeast extract) broth - 100 g - 1.01868.0100
D(+)-Glucose monohydrate - 1 kg Plastic bottle for solids 1.08342.1000
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty