Insulated shippers, STOROpack
Supplier: Sonoco
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STOROpack
216-1323EA 626 GBP
216-1323
Insulated shippers, STOROpack
Mailers and Shipping Containers
EPS (expanded polystyrene) and Neopor.
These moulded containers are designed for logistics applications in the pharmaceutical and biotechnology industries, where temperature-sensitive products have to be transported and stored in a temperature range of 2 to 8 °C, and require special protection from heat and cold.