May Grunwald's eosin-methylene blue, Q Path® stain for cytology
Supplier: VWR Q-Path Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Q path®
10047118.EA
85.6
GBP
10047118.
10047018.
May Grunwald's eosin-methylene blue, Q Path® stain for cytology
May Grunwald's eosin-methylene blue
Ready to use May & Grunwald for cytological staining in safe and easy to use pouch.
IVD registered. Instructions for use on vwr.com- just search for the product.
Storage Temperature:
Ambient |
MDL Number:
MFCD00131580 UN: 1170 ADR: 3,II |
Specification Test Results
Appearance | Clear liquid |
Colour of solution | Dark blue |
Density (20/4) | 0,79 - 0,81 |
pH (20°C) | 7 - 8,5 |
Absorbance (0.1 %) | 655 - 664 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...