4-(2-Chlorophenoxy)piperidine hydrochloride
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H51029.03EA
179
GBP
H51029.03
4-(2-Chlorophenoxy)piperidine hydrochloride
4-(2-Chlorophenoxy)piperidine hydrochloride
Formula:
C₁₁H₁₅Cl₂NO MW: 248.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02684107 CAS Number: 849107-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...