3-(2-Chloro-5-methylphenoxy)piperidine ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H50991.MDEA
52.2
GBP
H50991.MD
3-(2-Chloro-5-methylphenoxy)piperidine ≥98%
3-(2-Chloro-5-methylphenoxy)piperidine
Formula:
C₁₂H₁₆ClNO MW: 225.72 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08061103 CAS Number: 902837-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...