2,5-Dichlorothiobenzamide ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H32323.03EA
72.1
GBP
H32323.03
2,5-Dichlorothiobenzamide ≥97%
2,5-Dichlorothiobenzamide
Formula:
C₇H₅Cl₂NS MW: 206.09 g/mol Melting Pt: 88…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD09025686 CAS Number: 69622-81-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...