Methyl-3-cyano-5-fluorobenzoate 96%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Methyl 3-cyano-5-fluorobenzoate
H31759.MDEA
33.4
GBP
H31759.MD
Methyl-3-cyano-5-fluorobenzoate 96%
Methyl 3-cyano-5-fluorobenzoate
Formula:
C₉H₆FNO₂ MW: 179.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08688582 CAS Number: 886732-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...