3-Cyano-5-fluorophenylboronic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(3-Cyano-5-fluorophenyl)boronic acid
ACTIAS31238G5EA
191
GBP
ACTIAS31238G5
ACTIAS31238G1
3-Cyano-5-fluorophenylboronic acid
3-Cyano-5-fluorophenylboronic acid
Formula:
C₇H₅BFNO₂ MW: 164.9300 g/mol |
MDL Number:
MFCD09263608 CAS Number: 304858-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...