3,4-Dihydroxythiobenzamide ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H54875.03EA
61.9
GBP
H54875.03
3,4-Dihydroxythiobenzamide ≥97%
3,4-Dihydroxythiobenzamide
Formula:
C₇H₇NO₂S MW: 169.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09047665 CAS Number: 138038-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...