2-Carboxy-4-fluorophenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Borono-5-fluorobenzoic acid
COBBBB-3363-5GEA
118.71
GBP
COBBBB-3363-5G
COBBBB-3363-1G
2-Carboxy-4-fluorophenylboronic acid
2-Carboxy-4-fluorophenylboronic acid
Formula:
C₇H₆BFO₄ MW: 183.9296 g/mol |
MDL Number:
MFCD08235078 CAS Number: 874290-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...