Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Lidocaine hydrochloride

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"3"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (ALEXALX550251G005)
Supplier:  Enzo Life Sciences
Description:   LIDOCAINE . HYDROCHLORIDE . HYDRATE 1 * 5 g
UOM:  1 * 5 g
Catalog Number: (PROOBP214)
Supplier:  LGC Standards PROMOCHEM
Description:   LIDOCAINE (LIGNOCAINE) HYDROCHLORIDE 1 * 50 mg
UOM:  1 * 50 mg
Catalog Number: (PROOICRS0338)
Supplier:  LGC Standards PROMOCHEM
Description:   LIDOCAINE HYDROCHLORIDE 1 * 100 mg
UOM:  1 * 100 mg
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
1 - 3  of 3