Microplate readers, Vis absorbance, Biochrom EZ Read 800 Plus ELISA and Research

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

WAPR80-4002-02EA 8680 GBP
WAPR80-4002-02 WAPR80-4002-03
Microplate readers, Vis absorbance, Biochrom EZ Read 800 Plus ELISA and Research
The Biochrom EZ Read 800 Plus is a fast and accurate microplate reader that is an ideal choice to keep up with a heavy workload in a busy high-throughput laboratory. The EZ Read 800 Plus microplate readers can be operated via a user interface or PC-controlled using Galapagos software.

  • Fast plate reading: 96-well plate measurement in 5 seconds – suitable for high throughput applications
  • Single or dual wavelength measurement
  • Timed interval reading: Ideal for kinetic assays
  • Endpoint measurements
  • Four shaking modes: for complete mixing of solution

The EZ Read 800 Plus ELISA is a single-channel filter-based absorbance reader. Supplied with four filters for the most common ELISA substrates. It is robust and quick to set up, making it suitable for multi-level users. Calibration is performed prior to each reading ensuring accurate readings and results are exported to analysis software.

The EZ Read 800 Plus Research is supplied with filter sets for ELISA, protein assays and cell biology assays.

The EZ Read 800 Plus readers come with onboard software for instrument control, with the ability to store up to 120 methods and 100 plate results. The onboard software enables complete plate setup, qualitative, quantitative and kinetics, a range of curve-fit methods (including linear, cubic spline, point-to-point and 4 parameter).

Alternatively, readers can be operated using a PC and the supplied Galapagos software. Featuring ‘click and drag’ functionality, Galapagos software enables configuration of plate layouts, multiple assays per plate and multiple plates per analysis.

ELISA filter set: 405 nm (PNPP, ABTS); 492 nm (OPD); 450 nm (Slow, Turbo and Ultra TMB); 620 nm (Ref wavelength)
Protein assay filter set: 595 nm (Bradford); 562 nm (BCA)
Cell biology assay filter set: 570 nm (MTT); 492 nm (XTT); 620 nm (Turbidity cell counting)

Ordering information: For full details of additional filters, options and accessories including QC plate for instrument validation and spare lamps, please contact Avantor.

Delivery information: Supplied with Galapagos PC software (compatible with Windows® 7 and 8, and XP onwards), wavelength filters (as specified below), manual, power cord and USB cable. Additional filters are available from 400 to 750 nm, please contact Avantor for details.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR