Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rotary evaporators, Hei-VAP Advantage, digital

Supplier: Heidolph
HL models with hand lift, ML models with motor lift

For routine distillation applications and reproducible results.

  • Large digital 3,5" LCD display indicates rotation speed, heating bath temperature and vapour temperature
  • For limited budgets, the optional manual vacuum controller is available
  • For precise vacuum control and reproducible results, the optional digital vacuum controller is recommended
  • Automatic process timer turns off the evaporator at a pre-programmed time and removes evaporating flask from bath on motor lift model

Reduce process times by up to 30%, depending on the selected model and peripheral devices. The chemically resistant vacuum seal reduces the spare parts budget up to 75% with prolonged operational lifespan.

As standard, models are supplied with a detachable control panel with a 1.3 m cable. Therefore parameters can be changed outside of a closed fume hood, providing greater operator safety and ultimate process control. An optional remote control operating panel allows the user to step away from the evaporator and still monitor all process data increasing flexibility at work.

  • The vapour tube is easy to remove from the drive; a flask ejector feature allows the user to easily remove sticking evaporating flasks
  • Two independent safety circuits prevent overheating of the heating bath
  • In case of a power cut, all motor lift models will remove the flask from the heating bath preventing potential thermal damage to the evaporated solution or safety hazards
  • To protect the user from splashes and scalding, hot water and oil can be removed from the bath through a drain port
  • All heating baths feature non slip handles with safety grip for safe and simple bath installation and removal
Delivery information: Delivery information: Supplied with water/oil bath, 1 l evaporation flask (29/32), 1 l receiving flask (35/20) and condenser glassware set.
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local VWR sales office for guidance and details.

Delivery information: Supplied with water/oil bath, 1 l evaporation flask (29/32), 1 l receiving flask (35/20) and condenser glassware set.
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local VWR sales office for guidance and details.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Type Version Glassware Pk VWR Catalog Number Quantity
Hei-VAP Advantage HL, G1, UK-plug Diagonal condenser standard 1 531-0703
Hei-VAP Advantage HL, G1, UK-plug Diagonal condenser coated* 1 531-0704
Hei-VAP Advantage HL, G3, UK-plug Vertical condenser standard 1 531-0705
Hei-VAP Advantage HL, G3, UK-plug Vertical condenser coated* 1 531-0706
Hei-VAP Advantage HL, G5, UK-plug Condensate trap standard 1 531-0707
Hei-VAP Advantage HL, G5, UK-plug Condensate trap coated* 1 531-0708
Hei-VAP Advantage HL, G6, UK-plug Vertical condenser for reflux coated* 1 531-0710
Hei-VAP Advantage HL, G6, UK-plug Vertical condenser for reflux standard 1 531-0709
Hei-VAP Advantage ML, G1, UK-plug Diagonal condenser coated* 1 531-0712
Hei-VAP Advantage ML, G1, UK-plug Diagonal condenser standard 1 531-0711
Hei-VAP Advantage ML, G3, UK-plug Vertical condenser standard 1 531-0713
Hei-VAP Advantage ML, G3, UK-plug Vertical condenser coated* 1 531-0714
Hei-VAP Advantage ML, G5, UK-plug Condensate trap standard 1 531-0715
Hei-VAP Advantage ML, G5, UK-plug Condensate trap coated* 1 531-0716
Hei-VAP Advantage ML, G6, UK-plug Vertical condenser for reflux standard 1 531-0717
Hei-VAP Advantage ML, G6, UK-plug Vertical condenser for reflux coated* 1 531-0718
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Model Hei-VAP Advantage
Temperature range (°C) 20...210
Rotation speed (min-1) 20 - 280
Heating capacity (W) 1300
Bath capacity (l) 4,5
W×D×H (mm) 490×395×430
Protection class IP 20

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Glassware set, G1, complete, NS 29/32 1 531-9452
Glassware set, G3, complete, NS 29/32 1 HEID513-00300-00
Vacuum pump Rotavac Valve Tec 1 531-0457
Glassware set, G1B (coated), complete, NS 29/32 1 HEID513-00110-00
Glassware set, G3B (coated), complete, NS 29/32 1 HEID513-00310-00
Glassware set, G5, complete, NS 29/32 1 HEID513-00500-00
Glassware set, G5B (coated), complete, NS 29/32 1 HEID513-00510-00
Condenser, G1, for G1 glassware 1 HEID514-00100-00
Glassware set, G6, complete, NS 29/32 1 HEID513-00600-00
Condenser G1B, plastic coated, for G1 glassware 1 HEID514-00110-00
Glassware set, G6B (coated), complete, NS 29/32 1 HEID513-00610-00
Condenser G3B, plastic coated, for G3 glassware 1 HEID514-00310-00
Insert cold trap, for G5 glassware 1 HEID514-00501-00
Vapor tube, NS 29/32, for G6 glassware 1 HEID514-00000-05
Vacuum fitting NS 29/32, for G3/G6 glassware 1 HEID514-00001-00
Condenser, G6, for G6 glassware 1 531-0871
Upgrade kit, for upgrade from Advantage to Precision model (motor lift) 1 531-0737
Heating bath liquid, 5 l, up to 220 °C 5 l HEID569-00600-00
Spare clamping sleeve 1 HEID23-30-01-05-31
Spare vapor tube NS 29/32 (Clamping sleeve is not included) 1 531-0178
Protective hood 1 531-0992
Vacuum box, Hei-VAP 1 531-0735
Upgrade kit, for upgrade from Advantage to Precision model (hand lift) 1 531-0736
Tubing set for vacuum and water connection, 12 m (6,25 m vaccum tube and 6,25 m water tube) 1 531-3578
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty