The kuroGEL Midi Plus 15 Horizontal standard gel electrophoresis unit provides the most popular comb throughput and tray-size options, but with additional casting options.
- Multi channel pipette compatible combs, with a maximum 30-sample throughput, reduce gel loading time, while preparatory combs enable nucleic acids to be scaled-up for cloning
- Buffer recirculation ports can be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formation
- Coloured loading strips for easy well detection when loading
- UV-transparent acrylic casting tray allows the user to handle the gel on the transilluminator with minimum risk of exposure to hazardous ethidium bromide
- Side handles, for safe and easy transportation around the laboratory
Gel tank supplied with removable casting tray; 2×1,0 mm thick, 16-sample combs; coloured loading strips and buffer recirculation ports.
Combs for kuroGEL Midi Plus 15
Combs are colour coded and height adjustable, offering complete control over loading volume and well depth to a maximum 120-sample throughput. Four comb positions with 3,5 cm intervals along the tray for faster separation of multiple samples.
Comb slots: 4; comb thickness: 1,0, 1,5 or 2,0 mm; comb throughput: 10 to 30 samples.
Accessories for kuroGEL Midi Plus 15
Casting options: Casting gates with integral silicone seals effectively seal the tray without the need for tape, provided the silicone gasket faces outwards. 15 cm long Super-Seals offer total versatility in casting, allowing the gel length to be tailored to each user's personal requirements.
Coloured loading strips (optional) aid visualisation during gel loading. Available in three different colours for easy visualisation of samples mixed with dyes such as bromophenol and xylene cyanol. Each colour strip has an adhesive backing allowing it to be easily affixed and removed with each application.
Gel scoops (optional) are ideal for safe and easy transfer of gels from staining or destaining tanks to a transilluminator or similar surface.
*Multi channel pipette compatible.