Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Precision balances with draught shield, Science Education

Supplier: VWR Collection
VWR®
Competitively priced electronic balances for precise weighing (to 1 mg) with applications in the education sector. All models have a stainless steel top pan.

  • Easy to use balances with small footprint
  • Selectable measuring units and counting function
  • Round glass shield with top access for the 611-2600 and 611-2601
  • Conventional 'square' draught shield with sliding doors for 611-2610
  • Hook for under-balance weighing
  • Software for liquid and solid density determination requires optional density kit (please ask for details)

Note: All these balances are now supplied without an internal battery to meet recent transport regulations. An optional external rechargeable battery (611-8684) may be purchased for portable operation.

Certifications: EN 61326-1:2013, EN 61010-1: 2010, EN 61010-2-010:2014, EN 50581:2012

Accessories information: Optional Tablet System (611-8683) is suitable for use with models denoted with *.




 

ORDER

Expand All / Collapse All
Model Weighing cap. Readability Pan/platform size Pk VWR Catalog Number Quantity
SE 203 LR * 210 g 0,001 g Ø 110 mm 1 611-2600
SE 203 LW * 210 g 0,001 g Ø 110 mm 1 611-2610
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk For VWR Catalog Number Quantity
Ioniser panel, allows user to retrofit an integrated ioniser 1 VWR semi-micro, analytical or precision balances fitted with draftshields which have 3 sliding doors 611-3864
External rechargeable battery for portable operation 1 VWR balances 611-8684
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty