Microplate readers, Vis and UV/Vis absorbance, Biochrom EZ Read LED

Supplier: Biochrom
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


736-0861EA 9000 GBP
WAPR80-4002-10
Microplate readers, Vis and UV/Vis absorbance, Biochrom EZ Read LED
The Biochrom EZ Read LED is a PC-controlled, single channel, LED-based absorbance reader. The novel LED technology means that lamps rarely need changing compared to traditional halogen lamps.

  • Fast plate reading: 96-well plate measurement in 5 seconds – suitable for high throughput applications
  • Single or multiple wavelength measurement
  • Timed interval reading: Ideal for kinetic assays
  • Endpoint measurements
  • Four shaking modes: For complete mixing of solutions

Supplied with LEDs for the most common ELISA substrates. It is robust and quick to set up, making it suitable for multi-level users. The EZ Read LED UV version can read down to 340 nm, making it ideal for NADH/NADPH assays. Calibration is performed prior to each reading ensuring accurate readings.

The readers come with Galapagos PC software for instrument control and data analysis. Featuring 'click and drag' functionality, Galapagos software enables configuration of plate layouts, multiple assays per plate and multiple plates per analysis.

ELISA LED set: 405 nm (PNPP, ABTS); 492 nm (OPD); 450 nm (Slow, Turbo and Ultra TMB); 620 nm (Ref wavelength)

Ordering information: For full details of additional LEDs and spare lamps, please contact your local Avantor sales office.

Delivery information: Supplied with Galapagos PC software on CD and USB stick (compatible with Windows® 7 and 8, and XP onwards), LEDs (EZ Read LED: 405, 450, 492, 620 nm;  EZ Read LED UV: 340, 405, 450, 492, 620 nm), manual, power cord and USB cable. Additional LEDs are available from 400 to 900 nm, please contact Avantor for details.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR