Microplate readers, Vis absorbance, Biochrom EZ Read 800 ELISA and Research

Supplier: Biochrom
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

WAPR80-4002-00EA 7750 GBP
WAPR80-4002-00 WAPR80-4002-01
Microplate readers, Vis absorbance, Biochrom EZ Read 800 ELISA and Research
Microplate Readers
The Biochrom EZ Read 800 is a fast and accurate microplate reader that is an ideal choice to keep up with a heavy workload in a busy high-throughput laboratory.

  • Fast plate reading: 96-well plate measurement in 5 seconds – suitable for high throughput applications
  • Single or dual wavelength measurement
  • Timed interval reading: Ideal for kinetic assays
  • Endpoint measurements
  • Four shaking modes: for complete mixing of solutions

The EZ Read 800 ELISA is a PC-controlled, single-channel filter-based absorbance reader. Supplied with four filters for the most common ELISA substrates. It is robust and quick to set up, making it suitable for multi-level users. Calibration is performed prior to each reading ensuring accurate readings and results are exported to analysis software.

The EZ Read 800 Research is supplied with filter sets for ELISA, protein assays and cell biology assays.

The readers come with Galapagos PC software for instrument control and data analysis. Featuring 'click and drag' functionality, Galapagos software enables configuration of plate layouts, multiple assays per plate and multiple plates per analysis.

ELISA filter set: 405 nm (PNPP, ABTS); 492 nm (OPD); 450 nm (Slow, Turbo and Ultra TMB); 620 nm (Ref wavelength)
Protein assay filter set: 595 nm (Bradford); 562 nm (BCA)
Cell biology assay filter set: 570 nm (MTT); 492 nm (XTT); 620 nm (Turbidity cell counting)

Ordering information: For full details of additional filters, options and accessories including QC plate for instrument validation and spare lamps, please contact Avantor.

Delivery information: Supplied with Galapagos PC software (compatible with Windows® 7 and 8, and XP onwards), wavelength filters (as specified below), manual, power cord and USB cable. Additional filters are available from 400 to 750 nm, please contact Avantor for details.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR