Bed bath wipes, pre-saturated, Oasis™
Supplier: Vernacare
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Oasis™
VERORSC908EA 0 GBP
VERORSC908
Bed bath wipes, pre-saturated, Oasis™
Wipes Disposable Wipes and Towels Presaturated Wipes
These wipes offer a practical solution to patients and people with limited mobility who are unable to easily take a bath or shower. Each wipe is pre-impregnated with a combination of skin cleaners, designed to remove sweat, dirt, urine and faeces while moisturisers nourish and hydrate the skin.
  • Wipes can be warmed in the unopened pack in a microwave or the Oasis™ Warmer, can also be used cool straight from the pack for a fresher feel on warm days
  • Pre-impregnated with a mild surfactant formula, pH balanced for healthy skin and contains no alcohol or lanolin
  • Gentle cleansing formula is quick drying: there is no need for additional towels to dry the skin
  • Disposable: Minimise cross-contamination risks as there is no need for reusable bathing items such as soap and flannels which can harbour bacteria
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904