Charcoal activated 0,5-1,0 mm (18-35 mesh) for gas chromatography, Supelco®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Activated carbon
, Carbon activated
, Activated charcoal
M-Clarity™ quality level = MQ100
1.09624.0500EA
836
GBP
1.09624.0500
Charcoal activated 0,5-1,0 mm (18-35 mesh) for gas chromatography, Supelco®
Charcoal activated
Formula:
C MW: 12.01 g/mol Melting Pt: 3550 °C Density: 2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 ADR: 4.2,III REACH: 01-2119488894-16 Merck Index: 13,01818 |
Specification Test Results
Resolution | 4.5 - 11.0 |
Bulk density | 0.4 - 0.5 g/cm³ |
Zn (Zinc) | ≤ 0.002 % |
Particle size (0.5 - 1.0 mm) | ≥ 70.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...