A rotational liquid disinfectant containing surface active agents and a blend of quaternary ammonium compounds (QAC) and biguanide for environmental and surface disinfection.
- Suitable for product contact areas, walls and floors
- Filled in Grade A environment located within the Class B room
- Use on pre-cleaned surfaces
- For professional use only
Crystel Platinum is ideal for the disinfection of walls and floors, and is safe for use on most surfaces found within pharmaceutical production facilities.
All disinfectants are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
* Add one 100 ml concentrate to 5 litres of water to make a 2% solution.
Certifications: EN standards: EN 1276 - bactericidal activity; EN 1650 - fungicidal activity; EN 13697 bactericidal surface test.
Delivery information: Supplied with its own certificate of analysis, sterility validation, certificate of irradiation, and detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data).
Packaging: Available in 100 ml concentrate bottles*, ready to use 900 ml (trigger spray) and 5 litre bottles. Ready to use packs can be used immediately for spraying and mopping.
Caution: Use biocides safely. Always read the label and product information before use.