Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Exhaust filters with or without change indicator

Supplier: SCAT Europe
The exhaust filter can be installed on all SCAT waste systems. It cleans solvent vapors from the exhaust air and is available in different sizes. With a specific surface of 1200 m²/g, the multi-component granulate is the optimum filter media for nearly all solvent vapors. It is based on active charcoal and contains additional components which prevent sticking or clump formation and so prevents restriction of the filter performance. 99% of the volatile substances are captured here.

Exhaust filters with change indicator can be activated by pushing the button. The change indicator shows the elapsed service life clearly and is easy to observe.



 

ORDER

Expand All / Collapse All
Type Pk VWR Catalog Number Quantity
Exhaust filter, size S, for waste containers with a volume up to 5 liters, with change indicator and splash protection, service life of 3 months 1 554-2712
Exhaust filter, size S, for waste containers with a volume up to 5 liters, fire resistant, service life of 3 months 1 590-0874
Exhaust filter, size M, for waste containers with a volume up to 20 liters, with change indicator and splash protection, service life of 6 months 1 554-2713
Exhaust filter, size M, for waste containers with a volume up to 20 liters, fire resistant, service life of 6 months 1 590-0876
Economy pack of 4 exhaust filters, size S, for waste containers with a volume up to 5 liters, with change indicator and splash protection, service life of 3 months 4 590-0001
Economy pack of 3 exhaust filters, size M, for waste containers with a volume up to 20 liters, with change indicator and splash protection, service life of 6 months 3 590-0002
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Exhaust filters with splash guard L, 120000 m² filtering surface 1 590-0680
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping