Platinum standard solution, 1,000 mg/L Pt in 10% hydrochloric acid (from Pt) standard for AAS and ICP
Supplier: Perkin Elmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PERCN9300140EA 156 GBP
PERCN9300140
Platinum standard solution, 1,000 mg/L Pt in 10% hydrochloric acid (from Pt) standard for AAS and ICP