4'-Bromo-3'-chloro-2'-methylacetanilide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-(4-Bromo-3-chloro-2-methylphenyl)acetamide
COBBAN-1278-1GEA
27.7
GBP
COBBAN-1278-1G
COBBAN-1278-5G
4'-Bromo-3'-chloro-2'-methylacetanilide
4'-Bromo-3'-chloro-2'-methylacetanilide
Formula:
C₉H₉BrClNO MW: 262.5300 g/mol |
MDL Number:
MFCD08059123 CAS Number: 125328-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...