4-Acetoxy-3-methoxyphenylboronic acid pinacol ester, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL594474-5GEA
316
GBP
SIAL594474-5G
4-Acetoxy-3-methoxyphenylboronic acid pinacol ester, Sigma-Aldrich®
4-Acetoxy-3-methoxyphenylboronic acid pinacol ester
Formula:
C₁₅H₂₁BO₅ |
MDL Number:
MFCD03789257 CAS Number: 811841-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...