Thioacetazone
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Acetylaminobenzaldehyde thiosemicarbazone
PROOICRS0430EA
0
GBP
PROOICRS0430
Thioacetazone
Thioacetazone
Formula:
C₁₀H₁₂N₄OS |
CAS Number:
104-06-3 EINECS: 203-170-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...