trans-Chlordane
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
442599.EA
220
GBP
442599.
trans-Chlordane
trans-Chlordane
Formula:
C₁₀H₆Cl₈ |
CAS Number:
5103-74-2 EINECS: 225-826-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...