Precision pocket thermometers with PT1000 probe, TD 130 and TD 110
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
620-1929EA 61.9 GBP
VWRI620-1929
Precision pocket thermometers with PT1000 probe, TD 130 and TD 110
These ready to use thermometers are designed for highly accurate, quality temperature management. They can be used in air, gas, liquids and solids. They are suitable for pharmaceutical and biotechnology companies, food industry, government institutes and environment.
  • Low battery warning indicator
  • Calibratable: Offset and slope input
  • 'HOLD' function
  • All probes are permanently connected to device
  • Housing is made of impact resistant ABS
  • Battery life: approx. 200 operating hours
Delivery information: Supplied with 9 volt battery, probe and manual.