4-Iodonicotinonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Cyano-4-iodopyridine

ACTIAS12124G1EA 253 GBP
ACTIAS12124MG500 ACTIAS12124G5 ACTIAS12124G1
4-Iodonicotinonitrile
4-Iodonicotinonitrile
Formula: C₆H₃IN₂
MW: 230.01 g/mol
MDL Number: MFCD09037453
CAS Number: 490039-72-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR