4-Bromo-2-methylbenzaldehyde
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACTIAS34590G1EA
0
GBP
ACTIAS34590G1
ACTIAS34590G10
ACTIAS34590G5
4-Bromo-2-methylbenzaldehyde
4-Bromo-2-methylbenzaldehyde
Formula:
C₈H₇BrO |
MDL Number:
MFCD07787171 CAS Number: 24078-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...