3-Chlorostyrene

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: m-Chlorostyrene

ACTIAS24970G25EA 182 GBP
ACTIAS24970G25
3-Chlorostyrene
3-Chlorostyrene
Formula: C₈H₇Cl
MW: 138.6 g/mol
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: –26 °C
Density: 1,09 g/cm³ (20 °C)
MDL Number: MFCD00000598
CAS Number: 2039-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR