2,5-Dichlorobenzylamine
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
(2,5-Dichlorophenyl)methanamine
ACTIAS20505G25EA
297
GBP
ACTIAS20505G25
ACTIAS20505G5
ACTIAS20505G1
2,5-Dichlorobenzylamine
2,5-Dichlorobenzylamine
Formula:
C₇H₇Cl₂N MW: 176.04 g/mol Boiling Pt: 124…126 °C (13 mmHg) Density: 1.2334 g/cm³ (20 °C) |
MDL Number:
MFCD00052391 CAS Number: 10541-69-2 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...