2,3-Difluorobenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

ACTIAS24917G100EA 293 GBP
ACTIAS24917G100 ACTIAS24917G25 ACTIAS24917G5
2,3-Difluorobenzyl alcohol
2,3-Difluorobenzyl alcohol
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1,28 g/cm³ (20 °C)
MDL Number: MFCD00010312
CAS Number: 75853-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR