Potassium hydrogen diiodate, Supelco®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Potassium bi-iodate
M-Clarity™ quality level = MQ300
1.04867.0050EA
145
GBP
1.04867.0050
Potassium hydrogen diiodate, Supelco®
Potassium hydrogen diiodate
Formula:
HI₂KO₆ MW: 389.91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011400 CAS Number: 13455-24-8 EINECS: 236-650-9 UN: 3085 ADR: 5.1 (8),II |
Specification Test Results
Solubility | 13 g/l (15 °C) |
Molar Mass | 389.91 g/mol |
Bulk Density | 1750 kg/m3 |
pH | 1 - 2 (50 g/l, H2O, 20 °C) |
Hazard Class | 5.1 |
Hazard Class Description | Oxidizer |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...