4-tert-Butylphenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4-(tert-Butyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

COBBPN-2280-5GEA 0 GBP
COBBPN-2280-5G COBBPN-2280-1G
4-tert-Butylphenylboronic acid pinacol ester
4-tert-Butylphenylboronic acid pinacol ester
Formula: C₁₆H₂₅BO₂
MW: 260.1800 g/mol
MDL Number: MFCD05663843
CAS Number: 214360-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR