5-Chloro-2-nitrophenylboronic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-Chloro-2-nitrophenyl)boronic acid

COBBBB-5287-5GEA 0 GBP
COBBBB-5287-5G COBBBB-5287-1G
5-Chloro-2-nitrophenylboronic acid
5-Chloro-2-nitrophenylboronic acid
Formula: C₆H₅BClNO₄
MW: 201.3700 g/mol
MDL Number: MFCD10696660
CAS Number: 532924-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR